Documenti fondamentali
SYND191922
Synthware distilling receiver with hose connection
joint: ST/NS 19/22
Sinonimo/i:
Synthware D191922, glass distilling receiver
About This Item
Codice UNSPSC:
41104812
NACRES:
NB.51
Giunto
joint: ST/NS19/22
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware D191922
Categorie correlate
Descrizione generale
Cow-type design having three receivers spaced 40o apart. 105o angle between top outer joint and lower inner joints.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)