Documenti fondamentali
SYNA392440
Synthware transfer valve adapter
joint: ST/NS 24/40
Sinonimo/i:
Synthware A392440, transfer adapter with valve
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A392440
Categorie correlate
Descrizione generale
The 0-4 mm high vacuum valves have two Viton® O-rings on the shaft and an integral PTFE O-ring on the tip.
Note legali
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)