Z212644
Ace powder dispensing funnel
capacity 100 mL
Sinonimo/i:
funnel powder, glass funnel, glass funnels
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.43
Prodotti consigliati
Giunto
joint ST/NS24/40
Produttore/marchio commerciale
Ace Glass 723930
Capacità
100 mL
Descrizione generale
For powders and solids up to 25 mesh. Features a flexible PTFE screw thread wrapped on a precision PTFE-ceramic shaft contained within a precision bore housing. Completely inert materials allow flushing with solvents. No.15 Ace-Thred affords easy disassembly and cleaning.
Note: This funnel operates on a screw feed principal. The rate or amount of powder added is controlled by turning the knob which feeds the powder into the funnel like a conveyor.
Note: This funnel operates on a screw feed principal. The rate or amount of powder added is controlled by turning the knob which feeds the powder into the funnel like a conveyor.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)