SYNA922440
Synthware™ vacuum or inert gas adapter
male joint: ST/NS 24/40
Sinonimo/i:
Synthware A922440, glass vacuum adapter
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Prodotti consigliati
Giunto
male joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A922440
Descrizione generale
Synthware™ vacuum or gas inlet adapter. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection (10 mm O.D.) is at a 90 degree angle to the standard taper inner joint (24/40).
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)