Documenti fondamentali
SYNA921420
Synthware vacuum or inert gas adapter
male joint: ST/NS 14/20
Sinonimo/i:
Synthware 921420, glass vacuum adapter
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Giunto
male joint: ST/NS14/20
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A921420
Categorie correlate
Descrizione generale
Synthware vacuum or gas inlet adapter. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection (8 mm O.D.) is at a 90 degree angle to the standard taper inner joint (14/20).
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)