Z558575
Aldrich® microscale glassware kit
Economy
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Descrizione generale
Components:
- Z684139 Adapter, Claisen, 14/10, (1)
- Z684155 Adapter, Multipurpose, 14/10, (1)
- Z684244 Condenser, Jacketed Reflux, 14/10, (1)
- Z684252 Distillation Head, Hickman w/sideport, (1)
- Z684260 Flask, Round Bottom, 10 mL, 14/10, (1)
- Z684295 Spin Vane, 3 & 5 mL, (1)
- Z684341 Tube, Craig, Complete, 2 mL, (1)
- Z684368 Tube, Drying, 14/10, (1)
- Z684422 Vial, Conical, 3.0 mL, (1)
- Z684430 Vial, Conical, 5.0 mL, (1)
- Z684201 Plastic Kit Box, (1)
High-quality, low cost glassware kits designed to cover a broad range of organic reactions in student labs and chemical research. Kits contain microscale glassware needed to perform experiments described in the textbook Microscale Organic Laboratory. 14/10 ground-glass joints with holed-caps and O-ring seals provide secure, gas-tight connections.
Economy kit - supplied in a compartmentalized PP storage box with hinged lid. Will do 95% of experiments described in the textbook.
Deluxe kit - supplied in a foam-lined plastic carrying case. Covers all experiments in the textbook.
Economy kit - supplied in a compartmentalized PP storage box with hinged lid. Will do 95% of experiments described in the textbook.
Deluxe kit - supplied in a foam-lined plastic carrying case. Covers all experiments in the textbook.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Prodotti correlati
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z558575-1EA | 4061837636745 |