SYNA272419
Synthware™ anti-climb adapter
100 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 19/22
Sinonimo/i:
Synthware A272419, splash-guard adapter
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Prodotti consigliati
Giunto
bottom joint: ST/NS19/22
joint: ST/NS top24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A272419
Capacità
100 mL
Categorie correlate
Descrizione generale
Trap provides expanded volume for reactions that have a tendency to foam excessively. The inner vapor tube is sealed to the top of the The inner vapor tube is sealed to the top of thetrap to prevent entry into the condenser.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
SYNA272419-1EA | 4061832185194 |