Documenti fondamentali
SYND211100
Synthware vacuum jacketd Vigreux distillation head
joint: ST/NS 14/20, column H 100 mm, immersion depth 25 mm
Sinonimo/i:
Synthware D211100, short path head, Vigreux distilling head
About This Item
Codice UNSPSC:
41104812
NACRES:
NB.51
Giunto
joint: ST/NS14/20
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware D211100
Alt. colonna
100 mm
Prof. immersione
25 mm
Categorie correlate
Descrizione generale
Vacuum jacketed Vigreux distillation head having a sealed on condenser, braced for strength. Vacuum adapter has an inner joint and a sealed in drip tip. To be used with a 25 mm immersion thermometer
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)