SYND171014
Synthware™ Vigreux vacuum jacketed micro distillation head
joint: ST/NS 14/20, thermometer joint: ST/NS 10/18, immersion depth 25 mm
Sinonimo/i:
Synthware D171014, Short path head
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.51
Prodotti consigliati
Giunto
thermometer joint: ST/NS10/18
joint: ST/NS14/20
Produttore/marchio commerciale
D171014
Prof. immersione
25 mm
Categorie correlate
Descrizione generale
Synthware™ distillation head features a Vigreux column jacketed to improve distillation performance. The standard taper joint (10/18) accommodates a thermometer.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)