DWK73802-8425
KIMBLE® Solid Cap
black phenolic cap (with rubber liner), cap for 8-425
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Prodotti consigliati
Materiali
PTFE/rubber liner
black phenolic cap (with rubber liner)
Sterilità
non-sterile
Caratteristiche
class A
Confezionamento
case of 144 ea
Produttore/marchio commerciale
Kimble® 73802-8425
Raccordi
cap for 8-425
Descrizione generale
Class A Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
- Specially formulated phenolic cap material
- Autoclavable
- White rubber liners
- Excellent for general laboratory use
Caratteristiche e vantaggi
- Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
- PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
- Excellent for general laboratory use
- Identified in ASTM Specification E982, Class A requirements
Note legali
KIMBLE is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
DWK73802-8425 |