CLS8180100
Pyrex® Lifetime Red™ graduated oil centrifuge tube
capacity 100.0 mL, O.D. × L 38 mm × 200 mm
Sinonimo/i:
Corning® 8180
About This Item
Codice UNSPSC:
41121703
NACRES:
NB.43
Prodotti consigliati
Materiali
conical bottom tube
Confezionamento
case of 12
pack of 12
Produttore/marchio commerciale
Corning 8180-100
D.E. × lungh.
38 mm × 200 mm
Capacità
100.0 mL
Categorie correlate
Descrizione generale
In accordance with standard methods of testing for water and sediment in petroleum products. White graduations against a permanent red panel. Graduated upward to 0.5 mL in divisions of 0.05 mL; from 0.5 mL to 2 mL in divisions of 0.1 mL; from 2 mL to 3 mL in divisions of 0.2 mL; from 3 mL to 5 mL in divisions of 0.5 mL; from 5 mL to 10 mL in divisions of 1 mL; from 10 mL to 25 mL in divisions of 5 mL and at 50, 75 and 100 mL.
Note legali
Corning is a registered trademark of Corning, Inc.
Lifetime-Red is a trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
CLS8180100-1EA | |
CLS8180100-12EA | 4061831705232 |