Documenti fondamentali
BR778013
BRAND® centrifuge tube, beaded rim
volume 15 mL, conical bottom, graduated: no
Sinonimo/i:
Centrifuge tubes, Tubes
About This Item
Codice UNSPSC:
41121703
NACRES:
NB.11
Materiali
borosilicate glass
conical bottom
Caratteristiche
RCF 3,500 × g
graduated: no
Confezionamento
pack of 100
Produttore/marchio commerciale
BRAND 778013
Volume
15 mL
Descrizione generale
The Boro 3.3 Centrifuge Tube has a capacity of approximately 15 ml and features a robust wall thickness of 1.0 mm. Designed for high-performance applications, it can withstand relative centrifugal forces (RCF) of up to 3500. Ideal for laboratory use, this tube ensures durability and reliability during centrifugation, making it an essential tool for various scientific procedures.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)