Documenti fondamentali
BR701932
BRAND® replacement piston rod for TRANSFERPETTOR
for fixed and digital type 5-25, 20 and 25 μL
Sinonimo/i:
Accessories, Pipettes, Piston rod, Piston-operated pipettes, Positive displacement principle, Transferpettor
About This Item
Codice UNSPSC:
41121501
NACRES:
NB.01
Materiali
stainless steel
Confezionamento
pack of 3 ea
Produttore/marchio commerciale
BRAND 701932
Descrizione generale
The BRAND® Replacement Piston Rod for the TRANSFERPETTOR is designed to ensure optimal performance and precision in liquid handling. Starting from 20 µl, product comes equipped with a seal. The replacement rod is easy to install and maintain, providing a reliable solution for users seeking to extend the life of their TRANSFERPETTOR while maintaining accuracy and efficiency in their pipetting tasks.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)