BR139820
BRAND® double ended laboratory spatula/spoon
L 210 mm
Sinonimo/i:
Spatula
About This Item
Codice UNSPSC:
41122403
NACRES:
NB.35
Prodotti consigliati
Materiali
polystyrene
Produttore/marchio commerciale
BRAND 139820
Lunghezza
210 mm
Descrizione generale
The BRAND® Double-Ended Laboratory Spatula/Spoon is a versatile and essential tool designed for a variety of laboratory applications. Made from impact-resistant polystyrene (PS), this spatula/spoon combination is perfect for mixing, transferring, and handling different materials in both research and educational environments.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
BR139820-10EA | 4061831853278 |