BAB609011100
SP Bel-Art® Durac® Calibrated Thermocouple Thermometer
temp. range at -200-1370 °C, 4 PROBE (K/J/E/T)
Sinonimo/i:
H-B calibrated thermocouple thermometer for K/J/E/T type probes
About This Item
Codice UNSPSC:
41112303
NACRES:
NB.71
Prodotti consigliati
Caratteristiche
4 PROBE (K/J/E/T)
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
HB Instruments B60901-1100
Parametri
temp. range at -200-1370 °C
Categorie correlate
Descrizione generale
Used for temperature measurement in life science research, kilns, gas turbine exhaust, diesel engines, and other industrial processes, H-B Calibrated Thermocouple Thermometers provide measurement throughout a broad temperature range and the ability to utilize multiple probes at one time.
Caratteristiche e vantaggi
- Wide temperature range allows accurate monitoring at extreme temperatures
- Max/min/average temperature recording capability
- Data hold
- Includes two K-type beaded wire probes (-20/250°C capability); one is calibrated and serialized
- Software CD (where included) for real time data collection and logging via USB
- Bluetooth® app available for use with B60911-0200
- Probes for wider temperature ranges available (B60901-1200 & B60901-1300) or use alternatively purchased probes; see chart for probe types accepted by each unit
- Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST
- Individually serialized, CE-marked Note: calibration applies to included probe and tested port only
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Bluetooth is a registered trademark of Bluetooth SIG, Inc.
Durac is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)