Documentos Principais
57123
Supelco
SPE Tube Adapter
configured for AutoTrace Automated Systems, for use with 3mL tubes, pk of 6
About This Item
Código UNSPSC:
12000000
embalagem
pk of 6
Nível de qualidade
técnica(s)
solid phase extraction (SPE): suitable
aplicação(ões)
food and beverages
compatibilidade
configured for AutoTrace Automated Systems
for use with 3mL tubes
Descrição geral
Tube adapters serve many purposes. They can be used to stack one SPE tube on top of another to provide different selectivities. A larger empty syringe barrel can be stacked on top of a smaller SPE tube to act as a larger load reservoir. Or, they can serve as an adapter for positive pressure methods (e.g. from a syringe or air/ N2 line).
Escolha uma das versões mais recentes:
Certificados de análise (COA)