Documentos Principais
22021
Swagelok® Nut
Swagelok® 102-1, brass, 1/16 in. Swagelok
About This Item
Código UNSPSC:
41115717
Materiais
brass
embalagem
pkg of 10 ea
fabricante/nome comercial
Swagelok® 102-1
conector
1/16 in. Swagelok
compatibilidade
for use with general purpose Supeltex ferrules
Descrição geral
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.
Aplicação
Swagelok® nut, front and back ferrule set maybe used for sampler construction from a single capillary gas chromatographic column, for collection of semi-volatile organic compounds through a diffusion denuder.
Informações legais
Swagelok is a registered trademark of Swagelok Company
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Diffusion denuder for the collection of semivolatile organic compounds.
Krieger MS and Hites RA.
Environmental Science & Technology, 26 (8), 1551-1555 (1992)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)