Key Documents
22014
Swagelok® Nut, Front and Back Ferrule Set
Swagelok® 200-S, brass, 1/8 in. Swagelok
About This Item
Código UNSPSC:
41115717
Materiais
brass
embalagem
pkg of 10 ea
fabricante/nome comercial
Swagelok® 200-S
conector
1/8 in. Swagelok
Descrição geral
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. While assembling a compression tube fitting, the metal tubing should be inspected properly to ensure that the surface is smooth. Then slide on the nut with the open side facing the end of the tubing. Next, slide on the back ferrule with the wider part facing the nut. In the next step, slide on the front ferrule with the small end of the cone facing the end of the tubing. Then insert the tubing into the fitting and tighten up the nut/ferrule assembly onto the fitting.
Aplicação
Swagelok® nut, front and back ferrule set maybe used for sampler construction from a single capillary gas chromatographic column, for collection of semi-volatile organic compounds through a diffusion denuder.
Informações legais
Swagelok is a registered trademark of Swagelok Company
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Lot/Batch Number
Lamentamos, não temos COA para este produto disponíveis online no momento.
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 7
1 of 7
Swagelok® Fittings Kit
22668-U
Swagelok® Fittings Kit
Swagelok® Front Ferrule Swagelok® 203-1, stainless steel, 1/8 in. Swagelok
22038
Swagelok® Front Ferrule
PTFE Tubing tubing L × O.D. × I.D. 25 ft × 1/4 in. (6.35 mm) × 0.228 in. (5.8 mm)
Supelco
20533
PTFE Tubing
Swagelok® Union Swagelok® 400-6, brass, 1/4 in.
22004
Swagelok® Union
Swagelok® Back Ferrule Swagelok® 204-1, stainless steel, 1/8 in. Swagelok
22039
Swagelok® Back Ferrule
Diffusion denuder for the collection of semivolatile organic compounds.
Krieger MS and Hites RA.
Environmental Science & Technology, 26 (8), 1551-1555 (1992)
Grob RL & Barry EF.
Modern Practice of Gas Chromatography, 522-523 (2004)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)