Key Documents
Z380083
Sigma-Aldrich
Steri Sterilizer replacement glass beads
for Steri 350
About This Item
Código UNSPSC:
41102422
NACRES:
NB.22
peso
300 g (container)quantity of beads detemined by weight
Embalagem
Packages measured in grams determine the right quantity for each unit.
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 4
1 of 4
Solid-glass beads borosilicate, diam. 1 mm
Z273619
Solid-glass beads
Solid-glass beads borosilicate, diam. 2 mm
Z273627
Solid-glass beads
Glass beads diam. ~5 mm
Sigma-Aldrich
18406
Glass beads