Documentos Principais
Z327050
BRAND® BLAUBRAND® volumetric flask, glass stopper, amber glass
volume 50 mL, accuracy: 0.06 mL
About This Item
Código UNSPSC:
41121804
NACRES:
NB.43
Materiais
borosilicate glass
wide-neck
Junta
joint ST/NS neck14/23
esterilidade
non-sterile
Características
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
closure type Glass stopper
embalagem
pack of 2
fabricante/nome comercial
BRAND
faixa de medição de volume
50 mL, accuracy: 0.06 mL
capacidade
50 mL
Descrição geral
Volumetric flasks according to DIN EN ISO 1042, Class A, conformity certified. Made from Boro 3.3, calibrated to contain (TC, IN). Supplied with a interchangeable amber glass stopper. Marks and inscriptions in high contrast white enamel.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
Outras notas
BR37445 (India)
Informações legais
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Protocolos
HPLC separation of 17 important cannabinoids including CBD, delta 9 THC and THCA. Read the application note
This page contains detailed information on working with volumetric flasks and how to prepare a standard solution with a volumetric flask.