Z325473
BRAND® SILBERBRAND ETERNA graduated pipette, calibrated to deliver (TD, EX)
capacity 2 mL
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Produtos recomendados
Materiais
AR-Glass
Características
class B
graduation
subdivision 0.01 mL
supplementary to DIN range
embalagem
pack of 12
fabricante/nome comercial
BRAND 27072
capacidade
2 mL
Descrição geral
AR-Glas®. Graduated pipettes according to DIN EN ISO 835. Type 3 (zero point at the top), total delivery. Graduations and inscriptions in ETERNA amber stain, particularly resistant to aggressive cleaning methods.
Outras notas
BR27072 (India)
Informações legais
AR-Glas is a registered trademark of Schott AG
BRAND is a registered trademark of BRAND GMBH + CO KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)