Key Documents
F4642
Dressing tissue forceps
L 12 in.
Sinônimo(s):
grasping aid, tongs, tweezers
About This Item
Código UNSPSC:
41122405
NACRES:
NB.35
Materiais
stainless steel
embalagem
pack of 1 ea
Comprimento
12 in.
Descrição geral
with serrated tips
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 9
1 of 9
Stainless steel forceps Specimen, L 5 1/2 in.
Z225592
Stainless steel forceps
Stainless steel forceps Medium tip, straight
Z168750
Stainless steel forceps
Stainless steel forceps Specimen, L 8 in.
Z225606
Stainless steel forceps
Artis tweezer Style 5-SA
Z742676
Artis tweezer
Tweezers Style #7, sharp, hooked, non-magnetic alloy (tips may be restraightened when bent)
T4787
Tweezers
Stainless steel forceps Fine tip, curved
Z168785
Stainless steel forceps
Micro-dissecting forceps straight
F4017
Micro-dissecting forceps
Artis Tweezer Style 7-SA
Z742677
Artis Tweezer