AXYMTS1112C
MiniTube
12-strip, bulk pack, size 1.1 mL, pkg of 5x80strips/cs, non-sterile, polypropylene
Sinônimo(s):
centrifuge tubes
About This Item
Produtos recomendados
Materiais
polypropylene
esterilidade
non-sterile
fabricante/nome comercial
Axygen Inc MTS-11-12-C
Corning
embalagem
pkg of 5x80strips/cs
tamanho
1.1 mL
Características e benefícios
- Ensures positive sample identification due to one-way lid design, in conjunction with alpha-numeric grid
- DNase/RNase free and non-pyrogenic in nature
- Compatible with Tecan®, Beckman Coulter™, ANSI/SLAS 1-2004, microplates, and other robotic workstations
Informações legais
Corning is a registered trademark of Corning, Inc.
Coulter is a trademark of Beckman Coulter, Inc.
Tecan is a registered trademark of Tecan Group AG