Z678341
Finnpipette® F2 variable volume pipette
volume 100-1000 μL
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Produtos recomendados
Características
color-coded blue
electronic: no
single-channel
embalagem
pack of 1 ea
fabricante/nome comercial
Finn 4642090
volume
100-1000 μL
Categorias relacionadas
Descrição geral
Finnpipette F2 has a light pipetting action, is easy to use and is fully autoclavable. Accuracy and precision are the most vital aspect of any pipette so the Finnpipette F2 was developed with Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
- Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
- Big display with white numbers on black background for increased visibility
- Double action pipetting button with very light and smooth pipetting action
- Fully autoclavable
- Super blow-out piston with volumes of 50μl and below
Informações legais
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z678341-1EA | 4061833036426 |