Z678120
Finnpipette® F3 variable volume pipette
volume 10-100 μL
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Produtos recomendados
Características
color-coded yellow
electronic: no
single-channel
embalagem
pack of 1 ea
fabricante/nome comercial
Finn 4640040
volume
10-100 μL
Categorias relacionadas
Descrição geral
Finnpipette F3 has a double action pipetting button with rotating upper part enabling smooth and steady pipetting action at the same time as preventing accidental volume alterations. Finpipette F3 is equipped with the Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
- Double action pipetting button with very light and smooth pipetting action
- Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
- Big display with white numbers on black background for increased visibility
- Color coded for easy tip volume recognition
Informações legais
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z678120-1EA | 4061833036020 |