Z560758
Scienceware® fitting assortment kit
28 fittings to handle the most common laboratory connections
About This Item
Código UNSPSC:
41121815
NACRES:
NB.35
Produtos recomendados
embalagem
pkg of 1 ea
fabricante/nome comercial
Bel-Art F19626-0000
Descrição geral
28 fittings to handle the most common laboratory connections. Fittings can handle teperatures up to 135°C.
Contains:
Contains:
- 5 Straight connectors
- 3 Stepped fittings
- 8 Bifurcated ("Y") fittings
- 5 "T" connectors
- 5 Quick-disconnect fittings
- 2 Tubing clamps
Informações legais
Scienceware is a registered trademark of SP Scienceware, a division of SP
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z560758-1EA | 4061832265803 |