Z503088
Threaded vial connector
20-400 thread for 22-400
Sinônimo(s):
747205-2022
About This Item
Código UNSPSC:
41121815
NACRES:
NB.35
Produtos recomendados
conector
20-400 thread for 22-400
Descrição geral
These threaded connectors provide a convenient method to connect standard screw thread sample vials to glassware with GPI threads and to standard taper joint adapters.
• Excellent for collecting samples from rotary evaporator traps
• Made of glass filled nylon with inert PTFE inner seals
• Temp. range: up to 200 °C
• Excellent for collecting samples from rotary evaporator traps
• Made of glass filled nylon with inert PTFE inner seals
• Temp. range: up to 200 °C
Características e benefícios
- Connectors have PTFE seals and FKM O-Rings
- Blue glass-filled nylon
- For use to 150 °C
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z503088-1EA | 4061826729816 |