Z316520
Stuart™ Scientific Gyro rocker
STR9, speed 10-70 rpm
About This Item
Código UNSPSC:
41100000
Produtos recomendados
entrada CA/CC
230 V
grau
CE Compliant
Características
autoclavable
fabricante/nome comercial
STR9
disponibilidade
not available in USA and Canada
Parâmetros
10-70 rpm speed
plataforma C × L
360 mm × 360 mm , Fitted with non-slip mat
Descrição geral
Unit has a 3-D gyroscopic motion providing a gentle swirling movement which is ideally suited for mixing under low foaming conditions, including DNA extractions, staining and destaining gels, cell culture and general mixing applications.
- Optional 2 deck rack for platelet agitation
- Nylon coated steel, 2 decks
- Adjustable angle of tilt: 0 to 12 degrees.
Informações legais
Stuart is a trademark of Bibby Sterlin Ltd.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z316520GB-1EA | |
Z316520-1EA | |
Z316520CH-1EA | |
Z316520EU-1EA |