Documentos Principais
Z217085
Sigma-Aldrich
Scienceware® microtube cutter
for tubes up to 1.5 mL
About This Item
Código UNSPSC:
41121702
NACRES:
NB.11
fabricante/nome comercial
Bel-Art F21018-0000
Descrição geral
Cutters are perfect for retrieving radioactive pellets from the tips of tubes without resuspension or the risk of splattered radioactive substance. Works best with polypropylene and polyethylene tubes up to 1.5ml, not recommended for use with polystyrene tubes. Maximum cut diameter: 9.4mm (.37"). Replacement blades available.
Safely and accurately removes microcentrifuge tube tips
  • Eliminating the hazards of using knives or razor blades
Informações legais
Scienceware is a registered trademark of SP Scienceware, a division of SP
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 2
1 of 2
Straightening Pliers
58646
Straightening Pliers