Z189901
Mercury collector
H × O.D. 48 mm × 105 mm
About This Item
Código UNSPSC:
47131905
NACRES:
NB.35
Produtos recomendados
fabricante/nome comercial
Bel-Art
A × D.E.
48 mm × 105 mm
Descrição geral
Use to collect spilled mercury quickly, just unscrew the lid with the foam pad, and press pad firmly onto the spill. This pressure forces the mercury into the pad. When the lid is screwed back onto the jar, the pad is compressed against the jar’s freestanding perforated plate, releasing the mercury into the jar bottom.
Características e benefícios
- Container holds 125ml of mercury
- Use recommended disposal method
- Replacement pads should be affixed with rubber cement
- Dimensions: 10.5 x 4.8cm O.D. (4⅛ x 2") high
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z189901-1EA | 4061832257587 |