DWK45066C-18
KIMBLE® Solid Cap
black phenolic cap (with PTFE liner), cap for 18-415
About This Item
Código UNSPSC:
41121800
NACRES:
NB.11
Produtos recomendados
Materiais
PTFE/rubber liner
black phenolic cap (with PTFE liner)
esterilidade
non-sterile
Características
class B
embalagem
case of 225 ea
fabricante/nome comercial
Kimble® 45066C-18
conector
cap for 18-415
Categorias relacionadas
Descrição geral
Class B Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
- Specially formulated phenolic cap material
- Autoclavable
- White rubber liners
- Excellent for general laboratory use
Características e benefícios
- Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
- PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
- Excellent for general laboratory use
- Identified in ASTM Specification E982, Class A requirements
Informações legais
KIMBLE is a registered trademark of DWK Life Sciences
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
DWK45066C-18-225EA | 4061841117742 |