Documentos Principais
DWK45066C-15
KIMBLE® Solid Cap
black phenolic cap (with PTFE Liner), cap for 15-415
About This Item
Código UNSPSC:
41121800
NACRES:
NB.11
Materiais
PTFE/rubber liner
black phenolic cap (with PTFE Liner)
esterilidade
non-sterile
Características
class B
embalagem
case of 300 ea
fabricante/nome comercial
Kimble® 45066C-15
conector
cap for 15-415
Características e benefícios
  • Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirements
Informações legais
KIMBLE is a registered trademark of DWK Life Sciences
Escolha uma das versões mais recentes:
Certificados de análise (COA)