Key Documents
DWK212275408
DURAN® Super duty Erlenmeyer wide-neck flasks
flask capacity (1,000 mL)
Sinônimo(s):
212275408
About This Item
Código UNSPSC:
41121804
NACRES:
NB.17
Materiais
flat
Compatibilidade de esterilização
autoclavable compatible
Características
without cap
embalagem
case of 10 ea
fabricante/nome comercial
DURAN® 212275408
capacidade do frasco
(1,000 mL)
Descrição geral
Life can be tough for laboratory glassware such as beakers, flasks and cylinders. Rough handling and laboratory processes like automated washing can cause damage and breakages. The DURAN® SUPER DUTY products were specially developed to better cope with laboratory life. The DURAN® SUPER DUTY products are characterized by a higher mechanical strength achieved by reinforcing the rim. As a result of this modification, the impact strength is improved, and the risk of accidental breakage is significantly reduced. The DURAN® SUPER DUTY products increase the safety in the laboratory and help to minimize the risk of injury. The longer life expectancy of the SUPER DUTY products means that they are more economical to use over the long term.
Outras notas
Informações legais
DURAN is a registered trademark of DWK Life Sciences
Escolha uma das versões mais recentes:
Certificados de análise (COA)