Documentos Principais
CAD9926B
Animal feeding needle, sterile, disposable, PTFE w/SS bite protector
size 17G, L × diam. 3.3 in. × 2.4 mm , ball
About This Item
Código UNSPSC:
42142609
NACRES:
NB.03
tamanho
17G
embalagem
pkg of 20 x 5 ea (5 needles per sterile Tyvek peel pouch - 20 pouches per box)
C × diâmetro
3.3 in. × 2.4 mm , ball
Descrição geral
  • Flexible PTFE tube, covered at the hub end with type 304 stainless steel preventing tube damage due to biting during use
  • PTFE tube is softer than other tubing types, very flexible, and translucent for easy confirmation of reagent delivery
  • Plastic Luer hubs and soft silicone tips
  • Non-toxic and no-pyrogenic
  • Packaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box
Escolha uma das versões mais recentes:
Certificados de análise (COA)