Merck
EP5804736001
Eppendorf® Rotor Lid
aluminum, aerosol tight, for FA-45-30-11 ONLY (= 5804 726.006)
About This Item
UNSPSC-Code:
41103913
NACRES:
NB.82
Materialien
aluminum
Leistungsmerkmale
aerosol tight, for FA-45-30-11 ONLY (= 5804 726.006)
Hersteller/Markenname
Eppendorf® 5804736001
Methode(n)
centrifugation: suitable
Kompatibilität
for use with 5804 R
for use with 5804
for use with 5810 R
for use with 5810
Allgemeine Beschreibung
Rotor lids for Eppendorf centrifuge rotors.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG