Z743214
Julabo® Bad-Zubehör
installation cooing coil, for CORIO C and CD ranges, pack of 1 ea
Alle Fotos(1)
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.77
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
Julabo 9970101
Allgemeine Beschreibung
Add-on cooling coil (9 mm outer diameter) for counter-cooling with cooling water, can be used for tempering tasks close to the ambient temperature. A hose with an inner diameter of 10 mm is required. For use with Julabo® baths/circulators.
Rechtliche Hinweise
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z743214-1EA |