Merck
Z743212
Julabo® Bad-Zubehör
immersion-height adjustable platform, for CORIO C-B19/27 and CD-B19/27, pack of 1 ea
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.77
Verpackung
pack of 1 ea
Hersteller/Markenname
Julabo 9970502
Allgemeine Beschreibung
Stainles steel bath insert, variable shelf with adjustble height bath, for use with Julabo® CORIO C-B19/27 and CD-B19/27.
Rechtliche Hinweise
CORIO is a trademark of Julabo GmbH
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)