Merck
SYNF444000
Synthware three neck round bottom flask with vertical side necks
1000 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Synonym(e):
Synthware F444000, 3-neck round flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
round bottom
three-neck (vertical side necks)
Verbindung
center joint: ST/NS24/40
side joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F444000
Kapazität
1,000 mL
1000 mL
1000 mL
Allgemeine Beschreibung
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
SYNF444000-1EA4061832208619