Merck
SYNF644125
Synthware pressure equalizing funnel with high vacuum valve
125 mL, joint: ST/NS 24/40, valve size 0-4 mm
Synonym(e):
Synthware F644125, Addition funnel
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.51
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F644125
Kapazität
125 mL
125 mL
Verwandte Kategorien
Allgemeine Beschreibung
With PTFE extended tip valve for semi-needle valve adjustment.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)