SYND682440
Synthware™ distillation distribution adapter with vacuum hose connection
joint: ST/NS 24/40
Synonym(e):
Synthware D682440, radial style distillation receiver
Alle Fotos(1)
About This Item
UNSPSC-Code:
41104812
NACRES:
NB.51
Empfohlene Produkte
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware D682440
Verwandte Kategorien
Allgemeine Beschreibung
Cow-type design having three receivers spaced 40° apart. 105° angle between top outer joint and lower inner joints.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)