Merck
SYND681922
Synthware distillation distribution adapter with vacuum hose connection
joint: ST/NS 19/22
Synonym(e):
Synthware D681922, radial style distillation receiver
About This Item
UNSPSC-Code:
41104812
NACRES:
NB.51
Verbindung
joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware D681922
Verwandte Kategorien
Allgemeine Beschreibung
Cow-type design having three receivers spaced 40° apart. 105° angle between top outer joint and lower inner joints.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)