Merck
BAH500250546
SP Bel-Art® Glove Box Economy Sleeved Gloves for 8 in. Glove Ports
size 10 large
Synonym(e):
glove box
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.43
Materialien
black neoprene
Größe
10 large
8 in. (Glove port )
Verpackung
pkg of 1 pair
Hersteller/Markenname
Bel-Art H50025-0546
Verwandte Kategorien
Allgemeine Beschreibung
One piece, bonded neoprene protection against liquid or gas penetration.
Leistungsmerkmale und Vorteile
  • Use series 50029 for glove boxes with 6” glove ports (F50030-0000, F50028-2000)
  • Use series 50025 for glove boxes with 8” glove ports (H50026-0000)
  • Requires clamping rings to hold gloves in place
  • Sold in pairs
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)