Merck
Z108286
Tempo-Shield heat-resistant gloves
L 18 in.
About This Item
UNSPSC-Code:
46181504
NACRES:
NB.35
Materialien
Kevlar® (Asbestos-free heat-resistant gloves with wool liner)
Größe
one size fits all
Sterilität
non-sterile
Leistungsmerkmale
latex free
powder-free
reusable
Verpackung
pkg of 1 pair
Hersteller/Markenname
Sigma-Aldrich
Länge
18 in.
Allgemeine Beschreibung
Asbestos-free heat-resistant gloves with wool liner provide strength and heat protection up to 500 °F. Made from a blend of aluminized and synthetic fibers. Ambidextrous.
Rechtliche Hinweise
Kevlar is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)