Merck
Z677140
NevoCut cut protection gloves
size S
Synonym(e):
safety glove
About This Item
UNSPSC-Code:
46181504
NACRES:
NB.35
Materialien
white (polyamide, glass fibre and HPPE fibre steel)
white gloves
Größe
S
Sterilität
non-sterile
Leistungsmerkmale
latex free
powder-free
reusable
standard type EN388 (cat. II)
Verpackung
pack of 1 pair
Hersteller/Markenname
KCL 927
L × Dicke
250 mm × 1.50 mm
Allgemeine Beschreibung
Nevocut is the new generation of cut protection gloves combining good sensitivity, flexibility and comfort with the highest level of cut protection (level 5).
The combination of polyamide, glass fiber and high tensile PE fiber guarantees a high cut resistance without lowering the gloves dexterity.
Nevocut is ambidextrous and can be washed several times without loosing its excellent fit.
Rechtliche Hinweise
NevoCut is a trademark of KCL GmbH
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)