Z405299
Best® butyl gloves
size M
Alle Fotos(1)
About This Item
UNSPSC-Code:
46181504
NACRES:
NB.35
Empfohlene Produkte
Materialien
black gloves
butyl rubber
Größe
M
Sterilität
non-sterile
Leistungsmerkmale
latex free
powder-free
reusable
Verpackung
pkg of 1 pair
Hersteller/Markenname
Best Manufacturing 878-08
L × Dicke
350 mm × 0.70 mm
Anwendung
Best® butyl gloves are suitable for painting, coating preparation, chemical spray, and treatment. It can also be used for laboratory and pharmaceutical analysis.
Leistungsmerkmale und Vorteile
- Impermeable
- Resistant to water
- Great tactility and performance
- Provides permeation resistance to gases and water vapors
- Unsupported, heavy-duty, 100% butyl rubber
- Injection molded
- Smooth grip with rolled cuff
- Offers flexibility and durability
- Excellent resistance to ketones, acids, caustics, isocyanate, and gases
Rechtliche Hinweise
Best is a registered trademark of Showa Glove Co.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z405299-1PAK | 4061834378259 |