MilliporeSigma
HPLC Analysis of Phenolic Antioxidants on Ascentis® Express C18 2.7 μm
HPLC Analysis of Phenolic Antioxidants on Ascentis® Express C18 2.7 μm application for HPLC
CONDITIONS
column
Ascentis Express C18, 10 cm x 3.0 mm I.D., 2.7 μm partcles (53814-U)
mobile phase
[A] water; [B] acetonitrile; [C] 2-propanol;
gradient
Constant 43% C; 37 to 55% B in 3.6 min; held at 55% B for 2.4 min
flow rate
1 mL/min
pressure
4970 psi (343 bar)
column temp.
40 °C
detector
UV, 280 nm
injection
5 μL
sample
5 standards at 20 μg/mL
Description
Analysis Note
Phenolic Antioxidants are added to polyers for controlling their degradation process. Irgafos 168, Irganox 1010, Ethanox 330, Irganox 1076, BHT, and Irgafos 168 are often used as antioxidants to prevent the degradation of polypropylene polymer formulations. This HPLC method is for the quantitation of an additive or mix of additives in a polymeric material.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany