MilliporeSigma
HPLC Analysis of Atrazine Herbicides on Ascentis® Express C18
HPLC Analysis of Atrazine Herbicides on Ascentis® Express C18 application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 3 mm
Standard
Product No.
Description
Pricing
Atrazine
PESTANAL®, analytical standard
Isoproturon
PESTANAL®, analytical standard
Linuron
PESTANAL®, analytical standard
Diuron
PESTANAL®, analytical standard
Metazachlor
PESTANAL®, analytical standard
Methabenzthiazuron
PESTANAL®, analytical standard
Metobromuron
PESTANAL®, analytical standard
Metoxuron
PESTANAL®, analytical standard
Terbuthylazine-2-hydroxy
PESTANAL®, analytical standard
Monolinuron
PESTANAL®, analytical standard
Cyanazine
PESTANAL®, analytical standard
Hexazinone
PESTANAL®, analytical standard
CONDITIONS
column
Ascentis Express C18, 10 cm x 3.0 mm I.D., 2.7 μm particles (53814-U)
mobile phase
[A] 20 mM ammonium acetate, pH 6.4 unadjusted; [B] acetonitrile
gradient
20 to 28% B in 11 min; 28 to 65% B in 5 min; held at 65% B for 4 min
flow rate
0.6 mL/min
column temp.
46 °C
detector
UV, 240 nm
injection
5 μL
sample
10-25 μg/mL in 10:90, water: acetonitrile
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of 17 triazine pesticides.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany