US EPA Method 552: GC Analysis of Haloacetic Acids (Methyl Ester Derivatives) on Equity-1701
Materials
Analytical column
Product No.
Description
Pricing
CONDITIONS
column
Equity-1701, 30 m × 0.25 mm I.D., 0.25 mm (28372-U)
oven
40 °C (10 min.), 5 °C/min. to 75 °C (5 min.), 10 °C/min. to 150 °C (5min.)
inj. temp.
200 °C
detector
ECD, 300 °C
carrier gas
helium, 25 cm/sec @ 35 °C
injection
2 μL, splitless (0.5 min.)
liner
4 mm I.D. single taper
sample
12 component mix, 30-200 ppb in MTBE (47788, 46970-U and 47669-U)