MilliporeSigma
GC Analysis of Kerosene on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Kerosene on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis suitable for GC
Materials
Analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 10 m × 0.10 mm, df 0.10 μm
CONDITIONS
column
SLB-5ms, 10 m × 0.10 mm I.D., 0.10 μm (28465-U)
oven
40 °C, 80.0 °C/min to 150 °C, 70.0 °C/min to 250 °C, 50.0 °C/min to 320 °C
inj. temp.
320 °C
detector
FID, 320 °C
carrier gas
hydrogen, 85 cm/sec constant
injection
0.2 μL, 800:1 split
sample
kerosene
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany