Stable Isotopes
ISOTEC® Stable Isotopes are stable isotope compounds, ranging from gases to complex molecules. These compounds are useful for tracer studies in proteomics and metabolomics, agents for MRI / MRS, and in a wide range of other biomedical applications.
Both tracer and flux studies benefit from stable isotope label assistance. Metabolic pathways can be studied by tracking naturally-occurring and disease-indicated metabolites. Additionally, the effects of exogeneous substances or perturbations on the metabolome can be elucidated with the use of our stable isotope-labeled internal standards coupled with mass spectroscopy determination.
Products
Applications for Stable Isotopes
The use of our stable isotope-labeled compounds in metabolic and nutritional studies provides a means to dissect biochemical pathways. The availability of isotopically labeled compounds for basic scientific research permits the examination of the role of metabolic pathways in maintaining physiological states. In vivo studies using stable isotope labeled metabolites reveal important aspects of metabolism, including pathways and flux.
An important tool for high-resolution structure determination by NMR spectroscopy is the use of stable isotope-labeled biomolecules. By either selectively or uniformly incorporating stable isotopes into proteins, investigators can reduce the complexity of their spectra significantly. 13C, 15N, and deuterium are the most common isotopes incorporated into proteins through microbial expression systems, peptide synthesis, or with cell-free extracts.
The use of stable isotope compounds in medical imaging, in conjunction with 13C-MRS in vivo techniques, provides a method to explore fluxes through energy-related metabolic pathways for early diagnosis and treatment of neuropsychiatric disorders, cancer, and other diseases.
For these and many other applications, we offer a comprehensive portfolio of isotope labeled carbohydrates, amino acids, protected amino acids, metabolites, fatty acids, vitamins, steroids, other biologically-active compounds, and cell culture media and components. We also provide isotopically-labeled synthetic building blocks for researchers who prefer to make their own compounds. With our expertise in isotope synthesis, we stand ready to provide custom compounds for all of your specific needs.
Many of our stable isotope products are available from bulk stock. To receive pricing, availability and packaging information, complete the form below. You may also email your request to isosales@milliporesigma.com or contact the Stable Isotope Customer Service Team.
Fields indicated by an * are required.
Related Product Resources
Biomolecular NMR
Isotope Labeling Methods for Protein Dynamics Studies
Isotopic Labeling for NMR Spectroscopy of Biological Solids
Methyl Isotope-Labeling of Proteins
Utilizing ISOGRO® Supplementation of M9 Minimal Media
Selective Protonation of Methyl Groups in Highly Deuterated Proteins
Solid-state NMR on Larger Biomolecules
Application Note: a-Ketoacids as Biosynthetic Precursors for Protein NMR
Application Note: Isotopic Labeling in Solid-state NMR of Proteins
Application Note: Stable Isotope Labeling by Amino Acids in Cell Culture (SILAC)
Technical Bulletin: Utilizing ISOGRO® as Stand-alone or Supplemental Media
Mass Spec
Isotopically Labeled Steroid Standards
Application Note: Comparison of Deuterium, 13C, and 15N Isotopic Labels in Mass Spec Standards
MRI / MRS / Hyperpolarization / PET
Parahydrogen Induced Polarization
13C MRE to Probe Brain Metabolism Using Hyperpolarized Pyruvic Acid
Dynamic Nuclear Polarization
MRI/MRS: Use of Isotopes in Hyperpolarization
Application Note: The Essential Role of Enriched Stable Nuclides in Positron Emission Tomography (PET)
Metabolism Research
Stable Isotope Labeled Fatty Acids as Metabolic Tracers
Tools for Measuring Energy Expenditure
Application Note: An Approach to Quantifying the conversion of Carbohydrate to Fat in Humans
Brochure: Metabolomics
Highlights
13C labeled metabolites detection can render MRI images of metabolic changes in the body with the goal of diagnosis of cancer and other diseases.